Assay Details
| Gene Symbol: Kctd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109141.1 | NP_001102611.1 | 4 - 5 | 672 | 73 | |
| XM_006228859.2 | XP_006228921.1 | 5 - 6 | 523 | 73 | ||
| XM_006228860.2 | XP_006228922.1 | 5 - 6 | 776 | 73 | ||
| XM_008759178.1 | XP_008757400.1 | 5 - 6 | 598 | 73 | ||
| GenBank mRNA | BC168920.1 | - | 4 - 5 | 616 | 9100405 | 73 |
More Information
Updated Mapping Information
Set Membership: |
|

Kctd15
Genomic Map