Assay Details
| Gene Symbol: Serhl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130579.1 | NP_001124051.1 | 5 - 6 | 454 | 105 | |
| XM_008765775.1 | XP_008763997.1 | 5 - 6 | 440 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Serhl2
Genomic Map