Assay Details
| Gene Symbol: Ddit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109986.1 | NP_001103456.1 | 1 - 2 | 105 | 79 | |
| XM_006241444.2 | XP_006241506.1 | 2 - 3 | 1283 | 79 | ||
| GenBank mRNA | AW916370.1 | - | 1 - 2 | 105 | 79 | |
| BC100664.1 | - | 1 - 2 | 68 | 7932840 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC100664.1, NM_001109986.1, XM_006241444.2 transcripts.
Set Membership: |
|

Ddit3
Genomic Map