Assay Details
| Gene Symbol: Ddit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024134.2 | NP_077048.2 | 1 - 2 | 100 | 76 | |
| XM_006241445.2 | XP_006241507.1 | 2 - 3 | 1278 | 76 | ||
| GenBank mRNA | CF977695.1 | - | 1 - 2 | 103 | 76 | |
| U30186.1 | - | 1 - 2 | 92 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_024134.2, U30186.1, XM_006241445.2 transcripts.
Set Membership: |
|

Ddit3
Genomic Map