Assay Details
| Gene Symbol: Gcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024277.1 | NP_001019448.1 | 3 - 4 | 423 | 119 | |
| GenBank mRNA | BC092591.1 | - | 3 - 4 | 423 | 7372269 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|

Gcat
Genomic Map