Assay Details
| Gene Symbol: Fam83h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130565.1 | NP_001124037.1 | 3 - 4 | 761 | 64 | |
| GenBank mRNA | FQ222515.1 | - | 2 - 3 | 377 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam83h
Genomic Map