Assay Details
| Gene Symbol: Etv3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100992.2 | NP_001094462.2 | 4 | 640 | 80 | |
| XM_006232728.2 | XP_006232790.1 | 4 | 903 | 80 | ||
| XM_006232729.2 | XP_006232791.1 | 4 | 920 | 80 | ||
| XM_006232730.2 | XP_006232792.1 | 5 | 571 | 80 | ||
| XM_006232731.2 | XP_006232793.1 | 5 | 544 | 80 | ||
| GenBank mRNA | BC127525.1 | - | 4 | 640 | 7114610 | 80 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Etv3l
Genomic Map