Assay Details
| Gene Symbol: Trim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108552.1 | NP_001102022.1 | 5 - 6 | 986 | 72 | |
| GenBank mRNA | AY724524.1 | - | 5 - 6 | 971 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim2
Genomic Map