Assay Details
| Gene Symbol: S100a16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108557.2 | NP_001102027.1 | 2 - 3 | 222 | 73 | |
| XM_006232717.2 | XP_006232779.1 | 2 - 3 | 296 | 73 | ||
| XM_006232718.1 | XP_006232780.1 | 2 - 3 | 213 | 73 | ||
| GenBank mRNA | BC158598.1 | - | 2 - 3 | 222 | 7127572 | 73 |
| FQ209774.1 | - | 2 - 3 | 224 | 73 | ||
| FQ214291.1 | - | 2 - 3 | 205 | 73 | ||
| FQ214430.1 | - | 2 - 3 | 218 | 73 | ||
| FQ229313.1 | - | 2 - 3 | 230 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

S100a16
Genomic Map