Assay Details
| Gene Symbol: Mtx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100667.1 | NP_001094137.1 | 1 - 2 | 546 | 65 | |
| XM_006232612.2 | XP_006232674.1 | 1 - 2 | 1172 | 65 | ||
| XM_008761135.1 | XP_008759357.1 | 1 - 2 | 1184 | 65 | ||
| GenBank mRNA | BC099090.1 | - | 1 - 2 | 457 | 65 | |
| BC158632.1 | - | 1 - 2 | 546 | 7456627 | 65 | |
| FQ216274.1 | - | 1 - 2 | 437 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mtx1
Genomic Map