Assay Details
| Gene Symbol: Cd5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025685.1 | NP_001020856.1 | 4 - 5 | 771 | 98 | |
| GenBank mRNA | BC098724.1 | - | 4 - 5 | 771 | 7378264 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

Cd5l
Genomic Map