Assay Details
| Gene Symbol: Etnk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107894.1 | NP_001101364.1 | 7 - 8 | 1251 | 93 | |
| XM_006237623.1 | XP_006237685.1 | 7 - 8 | 1246 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Etnk1
Genomic Map