Assay Details
| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173386.1 | NP_001166857.1 | 1 - 2 | 144 | 67 | |
| XM_006237100.2 | XP_006237162.1 | 1 - 2 | 267 | 67 | ||
| GenBank mRNA | AB455106.1 | - | 1 - 2 | 236 | 67 | |
| AB455107.1 | - | 1 - 2 | 236 | 67 | ||
| BC166774.1 | - | 1 - 2 | 144 | 67 | ||
| GU357485.1 | - | 1 - 2 | 76 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clec7a
Genomic Map