Assay Details
| Gene Symbol: Med21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107895.2 | NP_001101365.2 | 1 - 2 | 72 | 88 | |
| GenBank mRNA | FM087109.1 | - | 1 - 2 | 72 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med21
Genomic Map