Assay Details
| Gene Symbol: LOC100911672 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750658.3 | XP_003750706.1 | 2 - 3 | 341 | 161 | |
| XM_006244637.2 | XP_006244699.1 | 2 - 3 | 327 | 161 | ||
| XM_008766913.1 | XP_008765135.1 | 2 - 3 | 309 | 161 | ||
| Gene Symbol: Atn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U31777.1 | - | 2 - 3 | 354 | 161 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the U31777.1,XM_003750658.3,XM_006244637.2,XM_008766913.1 transcripts.
This assay is in the 5'UTR region of the U31777.1,XM_003750658.3,XM_006244637.2,XM_008766913.1 transcripts.
Set Membership: |
|

LOC100911672
Genomic Map