Assay Details
Gene Symbol: LOC684471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750810.3 | XP_003750858.3 | 2 | 1908 | 66 | |
XM_008767723.1 | XP_008765945.1 | 2 | 2429 | 66 | ||
XM_008767724.1 | XP_008765946.1 | 3 | 1985 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
