Assay Details
| Gene Symbol: Spop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100496.1 | NP_001093966.1 | 3 - 4 | 361 | 96 | |
| XM_006247180.2 | XP_006247242.1 | 4 - 5 | 1165 | 96 | ||
| XM_006247181.2 | XP_006247243.1 | 5 - 6 | 472 | 96 | ||
| XM_006247182.2 | XP_006247244.1 | 4 - 5 | 421 | 96 | ||
| XM_008767968.1 | XP_008766190.1 | 4 - 5 | 517 | 96 | ||
| XM_008767969.1 | XP_008766191.1 | 5 - 6 | 439 | 96 | ||
| GenBank mRNA | BC091435.1 | - | 1 - 2 | 87 | 96 | |
| BC166743.1 | - | 3 - 4 | 361 | 7938440 | 96 | |
| FQ217100.1 | - | 5 - 6 | 669 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spop
Genomic Map