Assay Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031763.3 | NP_113951.1 | 896 | 98 | ||
| GenBank mRNA | BC072510.1 | - | 896 | 7104733 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pafah1b1
Genomic Map