Assay Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031763.3 | NP_113951.1 | 1504 | 66 | ||
| GenBank mRNA | AF016049.1 | - | 1319 | 66 | ||
| BC072510.1 | - | 1504 | 7104733 | 66 | ||
| FQ224997.1 | - | 1229 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Pafah1b1
Genomic Map