Assay Details
| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105796.2 | NP_001099266.2 | 18 - 19 | 2036 | 66 | |
| GenBank mRNA | FQ231544.1 | - | 2 - 3 | 293 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acap1
Genomic Map