Assay Details
| Gene Symbol: Nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037189.1 | NP_001032266.1 | 3 - 4 | 176 | 96 | |
| XM_008768018.1 | XP_008766240.1 | 2 - 3 | 1324 | 96 | ||
| GenBank mRNA | BC099120.1 | - | 3 - 4 | 176 | 7462721 | 96 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nsrp1
Genomic Map