Assay Details
| Gene Symbol: Nsrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037189.1 | NP_001032266.1 | 5 - 6 | 512 | 121 | |
| XM_008768018.1 | XP_008766240.1 | 4 - 5 | 1660 | 121 | ||
| GenBank mRNA | BC099120.1 | - | 5 - 6 | 512 | 7462721 | 121 |
More Information
Updated Mapping Information
Set Membership: |
|

Nsrp1
Genomic Map