Assay Details
| Gene Symbol: LOC103690090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773863.1 | XP_008772085.1 | 3 - 4 | 1020 | 147 | |
| XM_008773864.1 | XP_008772086.1 | 4 - 5 | 1038 | 147 | ||
| XM_008773865.1 | XP_008772087.1 | 3 - 4 | 252 | 147 | ||
| XM_008773866.1 | XP_008772088.1 | 2 - 3 | 167 | 147 | ||
| XM_008773867.1 | XP_008772089.1 | 3 - 4 | 184 | 147 | ||
| Gene Symbol: Ap2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080583.1 | NP_542150.1 | 3 - 4 | 281 | 147 | |
| XM_008767935.1 | XP_008766157.1 | 4 - 5 | 1038 | 147 | ||
| XM_008767936.1 | XP_008766158.1 | 3 - 4 | 252 | 147 | ||
| XM_008767937.1 | XP_008766159.1 | 2 - 3 | 167 | 147 | ||
| XM_008767938.1 | XP_008766160.1 | 2 - 3 | 184 | 147 | ||
| XM_008767939.1 | XP_008766161.1 | 3 - 4 | 1020 | 147 | ||
| GenBank mRNA | BC103481.1 | - | 4 - 5 | 250 | 7929959 | 147 |
| M77246.1 | - | 3 - 4 | 281 | 147 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690090
Genomic Map