Assay Details
| Gene Symbol: Krt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004022.2 | NP_001004022.1 | 6 - 7 | 1284 | 66 | |
| GenBank mRNA | BC101868.1 | - | 6 - 7 | 1284 | 7938504 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Krt15
Genomic Map