Assay Details
| Gene Symbol: Mbtd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768171.1 | XP_008766393.1 | 1 - 2 | 610 | 60 | |
| XM_008768172.1 | XP_008766394.1 | 1 - 2 | 610 | 60 | ||
| XM_008768173.1 | XP_008766395.1 | 1 - 2 | 610 | 60 | ||
| XM_008768174.1 | XP_008766396.1 | 1 - 2 | 610 | 60 | ||
| XM_008768175.1 | XP_008766397.1 | 3 - 4 | 308 | 60 | ||
| XM_008768176.1 | XP_008766398.1 | 2 - 3 | 247 | 60 | ||
| XM_008768177.1 | XP_008766399.1 | 2 - 3 | 522 | 60 | ||
| XM_008768178.1 | XP_008766400.1 | 2 - 3 | 260 | 60 | ||
| XM_008768179.1 | XP_008766401.1 | 2 - 3 | 395 | 60 | ||
| XM_008768180.1 | XP_008766402.1 | 1 - 2 | 199 | 60 | ||
| XM_008768181.1 | XP_008766403.1 | 1 - 2 | 195 | 60 | ||
| XM_008768182.1 | XP_008766404.1 | 1 - 2 | 610 | 60 | ||
| XM_008768183.1 | XP_008766405.1 | 4 - 5 | 469 | 60 | ||
More Information
Set Membership: |
|

Mbtd1
Genomic Map