Assay Details
| Gene Symbol: LOC100912585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246555.2 | XP_006246617.1 | 2 - 3 | 555 | 117 | |
| XM_006246556.2 | XP_006246618.1 | 2 - 3 | 1018 | 117 | ||
| XM_008767892.1 | XP_008766114.1 | 1 - 2 | 680 | 117 | ||
| Gene Symbol: Mapk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191547.1 | NP_001178476.1 | 236 | 117 | ||
| XM_006246417.2 | XP_006246479.1 | 555 | 117 | |||
| XM_006246418.2 | XP_006246480.1 | 1018 | 117 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100912585
Genomic Map