Assay Details
| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247274.2 | XP_006247336.1 | 1 - 2 | 143 | 107 | |
| XM_008767974.1 | XP_008766196.1 | 1 - 2 | 149 | 107 | ||
| GenBank mRNA | BC168163.1 | - | 1 - 2 | 86 | 8366401 | 107 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC168163.1, XM_006247274.2, XM_008767974.1 transcripts.
Set Membership: |
|

Nkiras2
Genomic Map