Assay Details
| Gene Symbol: Zfp692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006246383.2 | XP_006246445.1 | 1 - 2 | 476 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006246383.2 transcripts.
Set Membership: |
|

Zfp692
Genomic Map