Assay Details
| Gene Symbol: Zfp692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126272.1 | NP_001119744.1 | 3 - 4 | 793 | 69 | |
| XM_006246383.2 | XP_006246445.1 | 8 - 9 | 1504 | 69 | ||
| XM_006246384.2 | XP_006246446.1 | 8 - 9 | 1445 | 69 | ||
| XM_006246385.2 | XP_006246447.1 | 8 - 9 | 1192 | 69 | ||
| XM_006246386.2 | XP_006246448.1 | 8 - 9 | 1087 | 69 | ||
| GenBank mRNA | BC166433.1 | - | 8 - 9 | 1078 | 7191618 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp692
Genomic Map