Assay Details
| Gene Symbol: Scn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013178.1 | NP_037310.1 | 24 - 25 | 4714 | 93 | |
| XM_006247559.2 | XP_006247621.1 | 25 - 26 | 5064 | 93 | ||
| GenBank mRNA | M26643.1 | - | 24 - 25 | 4714 | 93 | |
| Y17153.1 | - | 23 - 24 | 4265 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24831 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083771 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083771 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083771 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P771870 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26552 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 617 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92839 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18413 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10748296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748297 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748299 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10748280 | - | - |
| Rat Genome 230 2.0 Array | 1387362_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381484_at | - | - |
| Rat Genome 230 2.0 Array | 1387362_at | - | - |
| Rat Genome 230 2.0 Array | 1381484_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387362_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387362_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000035928.rn.1 | TC1000003952.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387362_at | - | - |
| Rat Expression Set 230 | 1387362_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748280 | - | - |
| Rat Genome U34 Set | M26643_g_at | - | - |
| Rat Genome U34 Set | D87922exon_at | - | - |
| Rat Genome U34 Set | M26643_at | - | - |
| Rat Genome U34 Set | M26643_g_at | - | - |
| Rat Genome U34 Set | M26643_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21573053 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370344 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64597 | - | - |

Scn4a
Genomic Map