Assay Details
| Gene Symbol: LOC102552659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247245.2 | XP_006247307.2 | 5 - 6 | 591 | 144 | |
| Gene Symbol: Erbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017003.2 | NP_058699.2 | 18 - 19 | 2328 | 144 | |
| GenBank mRNA | AY116182.1 | - | 18 - 19 | 2328 | 144 | |
| BC061863.1 | - | 18 - 19 | 2400 | 5597835 | 144 | |
| X03362.1 | - | 18 - 19 | 2242 | 144 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102552659
Genomic Map