Assay Details
| Gene Symbol: Mrps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100471.1 | NP_001093941.1 | 4 - 5 | 518 | 92 | |
| GenBank mRNA | BC088232.1 | - | 4 - 5 | 507 | 92 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps7
Genomic Map