Assay Details
| Gene Symbol: Cnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012809.2 | NP_036941.1 | 2 - 3 | 753 | 106 | |
| XM_006247260.2 | XP_006247322.1 | 2 - 3 | 773 | 106 | ||
| XM_006247261.2 | XP_006247323.1 | 2 - 3 | 777 | 106 | ||
| XM_008767948.1 | XP_008766170.1 | 2 - 3 | 687 | 106 | ||
| GenBank mRNA | BC098066.1 | - | 2 - 3 | 753 | 7458826 | 106 |
| L16532.1 | - | 2 - 3 | 752 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnp
Genomic Map