Assay Details
| Gene Symbol: Msrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044285.2 | NP_001037750.2 | 2 - 3 | 276 | 71 | |
| GenBank mRNA | BC094309.1 | - | 2 - 3 | 266 | 7316479 | 71 |
| CB812976.1 | - | 2 - 3 | 276 | 71 | ||
| FQ220312.1 | - | 2 - 3 | 289 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msrb1
Genomic Map