Assay Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002727821.4 | XP_002727867.2 | 5 - 6 | 924 | 75 | |
| XM_006247473.2 | XP_006247535.1 | 5 - 6 | 924 | 75 | ||
| XM_006247474.2 | XP_006247536.1 | 4 - 5 | 1019 | 75 | ||
More Information
Set Membership: |
|

Fam171a2
Genomic Map