Assay Details
| Gene Symbol: Abca17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006245927.2 | XP_006245989.1 | 22 - 23 | 4773 | 63 | |
| XM_006245928.2 | XP_006245990.1 | 22 - 23 | 4757 | 63 | ||
| XM_008767554.1 | XP_008765776.1 | 22 - 23 | 5387 | 63 | ||
| XM_008767555.1 | XP_008765777.1 | 21 - 22 | 3760 | 63 | ||
| XM_008767556.1 | XP_008765778.1 | 21 - 22 | 3646 | 63 | ||
More Information
Set Membership: |
|

Abca17
Genomic Map