Assay Details
| Gene Symbol: Icam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006247627.2 | XP_006247689.1 | 1 - 2 | 192 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247627.2 transcripts.
Set Membership: |
|

Icam2
Genomic Map