Assay Details
| Gene Symbol: Eif4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100158.2 | NP_001093628.1 | 5 - 6 | 570 | 83 | |
| GenBank mRNA | BC105875.1 | - | 5 - 6 | 515 | 7130292 | 83 |
| CO559861.1 | - | 5 - 6 | 570 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif4a3
Genomic Map