Assay Details
| Gene Symbol: Gna13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013119.1 | NP_001013137.1 | 2 - 3 | 509 | 105 | |
| GenBank mRNA | AY553631.1 | - | 2 - 3 | 509 | 105 | |
| AY553632.1 | - | 2 - 3 | 509 | 105 | ||
| DQ120481.1 | - | 2 - 3 | 509 | 105 | ||
| DQ120482.1 | - | 2 - 3 | 509 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gna13
Genomic Map