Assay Details
| Gene Symbol: Ccdc154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767607.1 | XP_008765829.1 | 12 - 13 | 2110 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc154| Gene Symbol: Ccdc154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767607.1 | XP_008765829.1 | 12 - 13 | 2110 | 110 | |
Set Membership: |
|