Assay Details
| Gene Symbol: Havcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100762.1 | NP_001094232.1 | 3 - 4 | 552 | 98 | |
| GenBank mRNA | AJ549521.1 | - | 2 - 3 | 420 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Havcr2
Genomic Map