Assay Details
| Gene Symbol: Ccdc157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001062582.4 | XP_001062582.3 | 2 - 3 | 400 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc157| Gene Symbol: Ccdc157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001062582.4 | XP_001062582.3 | 2 - 3 | 400 | 83 | |
Set Membership: |
|