Assay Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107234.1 | NP_001100704.1 | 4 - 5 | 675 | 113 | |
| XM_006251300.1 | XP_006251362.1 | 4 - 5 | 701 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rnf215
Genomic Map