Assay Details
| Gene Symbol: Tnfsf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145681.1 | NP_663714.1 | 2 - 3 | 287 | 101 | |
| GenBank mRNA | AY115578.1 | - | 2 - 3 | 287 | 101 | |
| EF030546.1 | - | 2 - 3 | 278 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf10
Genomic Map