Assay Details
Gene Symbol: LOC499602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025039.2 | NP_001020210.2 | 2 - 3 | 160 | 142 | |
XM_006232310.2 | XP_006232372.1 | 2 - 3 | 380 | 142 | ||
GenBank mRNA | CK358190.1 | - | 2 - 3 | 218 | 142 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025039.2, XM_006232310.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
