Assay Details
| Gene Symbol: LOC499602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025039.2 | NP_001020210.2 | 5 - 6 | 426 | 99 | |
| XM_006232310.2 | XP_006232372.1 | 5 - 6 | 646 | 99 | ||
| XM_006232311.2 | XP_006232373.1 | 1 - 2 | 93 | 99 | ||
| XM_008760940.1 | XP_008759162.1 | 4 - 5 | 285 | 99 | ||
| GenBank mRNA | BC098046.1 | - | 1 - 2 | 65 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC499602
Genomic Map