Assay Details
| Gene Symbol: Pkia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053772.2 | NP_446224.1 | 3 - 4 | 346 | 82 | |
| GenBank mRNA | FM055544.1 | - | 3 - 4 | 351 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_053772.2 transcripts.
Set Membership: |
|

Pkia
Genomic Map