Assay Details
| Gene Symbol: Car3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019292.4 | NP_062165.2 | 1 - 2 | 41 | 148 | |
| XM_006232116.1 | XP_006232178.1 | 2 - 3 | 366 | 148 | ||
| GenBank mRNA | AB030829.1 | - | 1 - 2 | 41 | 148 | |
| AB030830.1 | - | 1 - 2 | 41 | 148 | ||
| AF037072.1 | - | 1 - 2 | 64 | 148 | ||
| BC061980.1 | - | 1 - 2 | 76 | 5597975 | 148 | |
| FQ210528.1 | - | 1 - 2 | 89 | 148 | ||
| FQ211459.1 | - | 1 - 2 | 88 | 148 | ||
| FQ214846.1 | - | 1 - 2 | 86 | 148 | ||
| FQ214929.1 | - | 1 - 2 | 91 | 148 | ||
| FQ216182.1 | - | 1 - 2 | 87 | 148 | ||
| FQ223846.1 | - | 1 - 2 | 87 | 148 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB030829.1,AB030830.1,AF037072.1,BC061980.1,NM_019292.4,XM_006232116.1 transcripts.
Set Membership: |
|

Car3
Genomic Map