Assay Details
| Gene Symbol: Fgf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019305.2 | NP_062178.1 | 2 - 3 | 810 | 76 | |
| GenBank mRNA | M22427.1 | - | 810 | 76 | ||
| X07285.1 | - | 2 - 3 | 531 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
angiogenesis
branching involved in ureteric bud morphogenesis
organ induction
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
phosphatidylinositol biosynthetic process
aging
positive regulation of cell proliferation
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
embryo development ending in birth or egg hatching
glial cell differentiation
positive regulation of endothelial cell migration
positive regulation of gene expression
negative regulation of fibroblast migration
positive regulation of phospholipase C activity
growth factor dependent regulation of skeletal muscle satellite cell proliferation
substantia nigra development
positive regulation of cerebellar granule cell precursor proliferation
hyaluronan catabolic process
lung development
inositol phosphate biosynthetic process
wound healing
positive regulation of cell fate specification
positive regulation of MAP kinase activity
positive regulation of blood vessel endothelial cell migration
negative regulation of blood vessel endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase activity
positive regulation of cell differentiation
positive regulation of osteoblast differentiation
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of retinal cell programmed cell death
positive regulation of smooth muscle cell proliferation
response to axon injury
stem cell development
positive regulation of epithelial cell proliferation
positive chemotaxis
release of sequestered calcium ion into cytosol
regulation of cell cycle
positive regulation of cell division
positive regulation of protein kinase B signaling
positive regulation of cardiac muscle cell proliferation
corticotropin hormone secreting cell differentiation
thyroid-stimulating hormone-secreting cell differentiation
negative regulation of cell death
chondroblast differentiation
mammary gland epithelial cell differentiation
angiogenesis involved in coronary vascular morphogenesis
negative regulation of wound healing
positive regulation of ERK1 and ERK2 cascade
positive regulation of canonical Wnt signaling pathway
positive regulation of sprouting angiogenesis
regulation of endothelial cell chemotaxis to fibroblast growth factor
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
fibroblast growth factor receptor binding
cytokine activity
protein binding
growth factor activity
heparin binding
ligand-dependent nuclear receptor transcription coactivator activity
chemoattractant activity
activation of MAPKK activity
angiogenesis
branching involved in ureteric bud morphogenesis
organ induction
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
phosphatidylinositol biosynthetic process
aging
positive regulation of cell proliferation
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
embryo development ending in birth or egg hatching
glial cell differentiation
positive regulation of endothelial cell migration
positive regulation of gene expression
negative regulation of fibroblast migration
positive regulation of phospholipase C activity
growth factor dependent regulation of skeletal muscle satellite cell proliferation
substantia nigra development
positive regulation of cerebellar granule cell precursor proliferation
hyaluronan catabolic process
lung development
inositol phosphate biosynthetic process
wound healing
positive regulation of cell fate specification
positive regulation of MAP kinase activity
positive regulation of blood vessel endothelial cell migration
negative regulation of blood vessel endothelial cell migration
positive regulation of phosphatidylinositol 3-kinase activity
positive regulation of cell differentiation
positive regulation of osteoblast differentiation
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulation of retinal cell programmed cell death
positive regulation of smooth muscle cell proliferation
response to axon injury
stem cell development
positive regulation of epithelial cell proliferation
positive chemotaxis
release of sequestered calcium ion into cytosol
regulation of cell cycle
positive regulation of cell division
positive regulation of protein kinase B signaling
positive regulation of cardiac muscle cell proliferation
corticotropin hormone secreting cell differentiation
thyroid-stimulating hormone-secreting cell differentiation
negative regulation of cell death
chondroblast differentiation
mammary gland epithelial cell differentiation
angiogenesis involved in coronary vascular morphogenesis
negative regulation of wound healing
positive regulation of ERK1 and ERK2 cascade
positive regulation of canonical Wnt signaling pathway
positive regulation of sprouting angiogenesis
regulation of endothelial cell chemotaxis to fibroblast growth factor
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
fibroblast growth factor receptor binding
cytokine activity
protein binding
growth factor activity
heparin binding
ligand-dependent nuclear receptor transcription coactivator activity
chemoattractant activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P212850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125276 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P212850 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P212850 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P212850 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P212850 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12014 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38086 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65421 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56387 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28603 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10815031 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815027 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815029 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10822930 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815030 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10822929 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10815026 | - | - |
| Rat Genome 230 2.0 Array | 1387606_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387606_at | - | - |
| Affymetrix HT Rat Focus Array | 1387606_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387606_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200006415.rn.1 | TC0200001019.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387606_PM_at | - | - |
| Rat Expression Set 230 | 1387606_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10815026 | - | - |
| Rat Genome U34 Set | M22427_at | - | - |
| Rat Genome U34 Set | X07285cds_s_at | - | - |
| Rat Genome U34 Set | X07285cds_s_at | - | - |
| Rat Genome U34 Set | M22427_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350498 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69660 | - | - |

Fgf2
Genomic Map