Assay Details
| Gene Symbol: Car2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019291.1 | NP_062164.1 | 3 - 4 | 355 | 73 | |
| GenBank mRNA | BC065577.1 | - | 3 - 4 | 388 | 5623601 | 73 |
| FQ212832.1 | - | 3 - 4 | 407 | 73 | ||
| FQ213417.1 | - | 3 - 4 | 407 | 73 | ||
| FQ213440.1 | - | 3 - 4 | 407 | 73 | ||
| FQ225794.1 | - | 3 - 4 | 405 | 73 | ||
| FQ225882.1 | - | 3 - 4 | 405 | 73 | ||
| FQ226264.1 | - | 3 - 4 | 407 | 73 | ||
| FQ228123.1 | - | 3 - 4 | 406 | 73 | ||
| FQ231140.1 | - | 3 - 4 | 405 | 73 | ||
| FQ232067.1 | - | 3 - 4 | 404 | 71 | ||
| FQ233035.1 | - | 3 - 4 | 406 | 72 | ||
| FQ233398.1 | - | 3 - 4 | 405 | 73 | ||
| FQ234177.1 | - | 3 - 4 | 404 | 72 | ||
| FQ234222.1 | - | 3 - 4 | 405 | 73 | ||
| FQ234668.1 | - | 3 - 4 | 407 | 73 | ||
| X58294.1 | - | 3 - 4 | 355 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car2
Genomic Map